Learn More
MSC Devcon 14260 50 mL Cartridge Two Part Epoxy 8 to 12 min Working Time, 2,250 psi Shear Strength
Catalog No. 501149321
Encompass
Devcon 14260 50 mL Cartridge Two Part Epoxy 8 to 12 min Working Time, 2,250 psi Shear Strength
Catalog No. 50-114-9321 Supplier MSC Supplier No. 00470799
Devcon 14260 50 mL Cartridge Two Part Epoxy 8 to 12 min Working Time, 2,250 psi Shear Strength