Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(Tributylstannyl)Thiophene

Catalog No. 41449250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-449-250ML 50 mL
41-449-210ML 10 mL
41-449-2250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 41-449-250ML Supplier Sigma Aldrich Supplier No. 41449250ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-(Tributylstannyl)thiophene has been used to synthesize diphenylquinoxaline monomer for the electrochromism polymers. It can also be used as a reactant in Stille coupling reaction.

Specifications

CAS 54663-78-4
Density 1.175 g/mL at 25°C
Boiling Point 155°C (0.1 mmHg, lit.)
Molecular Formula C16H30SSn
Refractive Index n20/D 1.518
Linear Formula C16H30SSn
MDL Number MFCD00192513
Quantity 50 mL
Molecular Weight (g/mol) 373.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.