Learn More
Cayman Chemical 2-Trifluoromethyl-2'-methoxychalcone, Purity: ≥98%, CAS: 1309371-03-6, 50 mg
SDP
Catalog No. 502952037
Encompass_Preferred
A potent activator of Nrf2; increases the expression of GCLM and NQO1 in human bronchial epithelial cells at 10 μM; increases GCLM and NQO1 expression by 4.5- and 4.6-fold, respectively, in the small intestine in mice at 50 mg/kg
Catalog No. 50-295-2037 Supplier Cayman Chemical Supplier No. 1188150 MG
A potent activator of Nrf2; increases the expression of GCLM and NQO1 in human bronchial epithelial cells at 10 μM; increases GCLM and NQO1 expression by 4.5- and 4.6-fold, respectively, in the small intestine in mice at 50 mg/kg