Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex 2-Way Direct Lift Solenoid Valve; normally closed 1/16 12VDC
Catalog No. NC0575588
Encompass
Rated for 26" Hg vacuum. Maximum fluid temperature is 158¿F (70¿C). All valves have 15" lead wires. Valves are compact and lightweight ¿ mount them almost anywhere. Valves with a 1/16" orifice have mounting holes at the bottom only and valves with a 3/32" and 5/32" orifice have mounting holes at the top and bottom.
Catalog No. NC0575588 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0154001
Rated for 26" Hg vacuum. Maximum fluid temperature is 158¿F (70¿C). All valves have 15" lead wires. Valves are compact and lightweight ¿ mount them almost anywhere. Valves with a 1/16" orifice have mounting holes at the bottom only and valves with a 3/32" and 5/32" orifice have mounting holes at the top and bottom.