Learn More
Ibidi USA -Slide 2 Well Ph+, 15pk
SDP
Catalog No. NC1916238
Encompass
A chambered coverslip with 2 wells and a special intermediate plate for excellent phase contrast and high-end fluorescence microscopy. Excellent phase contrast over the entire well with no meniscus. All-in-one chamber slide used for cell culture and microscopy. Observation of the sample through the coverslip like bottom using high resolution micrscopy. Fast and easy immunofluorescence with no coverslips and no leakage. Surface Modification: ibiTreat: #1.5 polymer coverslip, tissue culture treated, sterilizedPcs./Box: 15 (individually packed)
This item is not returnable. View return policy
Catalog No. NC1916238 Supplier Ibidi USA Supplier No. 80296
A chambered coverslip with 2 wells and a special intermediate plate for excellent phase contrast and high-end fluorescence microscopy. Excellent phase contrast over the entire well with no meniscus. All-in-one chamber slide used for cell culture and microscopy. Observation of the sample through the coverslip like bottom using high resolution micrscopy. Fast and easy immunofluorescence with no coverslips and no leakage. Surface Modification: ibiTreat: #1.5 polymer coverslip, tissue culture treated, sterilizedPcs./Box: 15 (individually packed)