Learn More
Apexbio Technology LLC 2X HyperFusion High-Fidelity Master Mix 10X1ml
SDP
Catalog No. 501998275
Encompass_Preferred
2X HyperFusion High-Fidelity Master Mix provides high fidelity strong amplification capability and faster amplification speed for most PCR applications HyperFusion DNA polymerase is composed of a Pyrococcus-like proofreading polymerase fused with a DNA-binding domain Its unique structure which combines a novel Pyrococcus-like enzyme and a domain enhancing synthesis capability increases fidelity and speed Due to its high fidelity HyperFusion DNA polymerase is an ideal choice for molecular cloning or other downstream applications HyperFusion DNA polymerase is among the DNA polymerases with the highest fidelity having a 50-fold lower error rate compared to Taq DNA polymerase and a 6-fold lower error rate than Pyrococcus Furious DNA polymerase HyperFusion DNA polymerase exhibits 5 to 3 polymerase activity and 3 to 5 exonuclease activity It generates blunt-ended PCR products without A-overhangs The extension rate is approximately 15-30 sec per kb (up to 10 kb in length)
Catalog No. 50-199-8275 Supplier Apexbio Technology LLC Supplier No. K103910
2X HyperFusion High-Fidelity Master Mix provides high fidelity strong amplification capability and faster amplification speed for most PCR applications HyperFusion DNA polymerase is composed of a Pyrococcus-like proofreading polymerase fused with a DNA-binding domain Its unique structure which combines a novel Pyrococcus-like enzyme and a domain enhancing synthesis capability increases fidelity and speed Due to its high fidelity HyperFusion DNA polymerase is an ideal choice for molecular cloning or other downstream applications HyperFusion DNA polymerase is among the DNA polymerases with the highest fidelity having a 50-fold lower error rate compared to Taq DNA polymerase and a 6-fold lower error rate than Pyrococcus Furious DNA polymerase HyperFusion DNA polymerase exhibits 5 to 3 polymerase activity and 3 to 5 exonuclease activity It generates blunt-ended PCR products without A-overhangs The extension rate is approximately 15-30 sec per kb (up to 10 kb in length)