Learn More
Apexbio Technology LLC 2HyPerFUsion High-Fidelity Master Mix (With dye), 1ml
SDP
Catalog No. 501998270
Encompass_Preferred
Our product 2HyPerFUsion High-Fidelity Master Mix (With dye) offers superior performance for major PCR applications with high fidelity, powerful amplification ability and faster amplification speed. HyPerFUsion high-fidelity DNA polymerases are composed of a DNA-binding domain fused to a Pyrococcus-like proofreading polymerase. Its unique structure, a novel Pyrococcus-like enzyme fused with a processivity-enhancing domain, increases fidelity and speed. The high fidelity makes hyPerFUsion high-fidelity DNA polymerase a superior choice for detection or other subsequent applications. HyPerFUsion high-fidelity DNA polymerase possesses one of the most accuracy with an error rate that is 50-fold lower compared to Taq DNA Polymerase and 6-fold lower than Pyrococcus Furiosus DNA Polymerase (Pfu). Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8270 Supplier Apexbio Technology LLC Supplier No. K10301
Our product 2HyPerFUsion High-Fidelity Master Mix (With dye) offers superior performance for major PCR applications with high fidelity, powerful amplification ability and faster amplification speed. HyPerFUsion high-fidelity DNA polymerases are composed of a DNA-binding domain fused to a Pyrococcus-like proofreading polymerase. Its unique structure, a novel Pyrococcus-like enzyme fused with a processivity-enhancing domain, increases fidelity and speed. The high fidelity makes hyPerFUsion high-fidelity DNA polymerase a superior choice for detection or other subsequent applications. HyPerFUsion high-fidelity DNA polymerase possesses one of the most accuracy with an error rate that is 50-fold lower compared to Taq DNA Polymerase and 6-fold lower than Pyrococcus Furiosus DNA Polymerase (Pfu). Other sizes are also available. Please inqury us for quote.