Learn More
Biolegend Brilliant Violet 421™ anti-human FOXP3
Supplier:  Biolegend 320124
Encompass_Preferred
Brilliant Violet 421™ anti-human FOXP3 [206D]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus, Baboon, Pigtailed Macaque; Apps: ICFC; Size: 100 tests
Catalog No. NC1076905