Learn More
IONBENCH CORPORATION 2X BANK OF THREE DRAWERS
Catalog No. NC3345426
Encompass_Preferred
  • Drawer module designed for laboratory workstations
  • Optimized for mass spectrometry (MS) and liquid chromatography (LC) environments
  • Two banks with three full-extension drawers each
  • Total of six drawers for optimal storage
  • Enhances organizational capacity for reagents and consumables
  • Seamlessly integrates into modular ionBench system
  • Contributes to improved workflow efficiency
  • Supports improved laboratory ergonomics
Catalog No. NC3345426 Supplier IONBENCH CORPORATION Supplier No. DR3SET
  • Drawer module designed for laboratory workstations
  • Optimized for mass spectrometry (MS) and liquid chromatography (LC) environments
  • Two banks with three full-extension drawers each
  • Total of six drawers for optimal storage
  • Enhances organizational capacity for reagents and consumables
  • Seamlessly integrates into modular ionBench system
  • Contributes to improved workflow efficiency
  • Supports improved laboratory ergonomics