Learn More
LGC GENOMICS LLC (Lucigen) 2X BHQ Probe Master Mix, Low ROX, 1.25mLA versatile mix for both end-point PCR genotyping and qPCR applications.
Catalog No. 502231945
Encompass
2X BHQ Probe Master Mix, Low ROX, 1.25mLA versatile mix for both end-point PCR genotyping and qPCR applications.
This item is not returnable. View return policy
Catalog No. 50-223-1945 Supplier LGC GENOMICS LLC Supplier No. KBS1040111
2X BHQ Probe Master Mix, Low ROX, 1.25mLA versatile mix for both end-point PCR genotyping and qPCR applications.