Learn More
101 BIO 2x Es Taq Master Mix (with dye) (1 mL, 100 rxn, 20 ul/rxn)
Catalog No. NC3486594
Encompass
This Master Mix contains Es Taq DNA Polymerase, PCR buffer, Mg2+, dNTP, PCR stabilizer and PCR enhancer. The concentration is 2x.Es Taq DNA Polymerase possesses 5´→3´ DNA polymerase and 5´→3´ exonuclease activity. The polymerase has the high amplification efficiency and low mismatching as Taq DNA polymerase and high fidelity of Pfu DNA polymerase. Es Taq Polymerase catalyzes the non-template directed addition of an adenine residue to the 3´-end of both strands of DNA molecules to make it suitable for T/A cloning. The amplification range of Es Taq is ~ 6 kb.This unique Master Mix recipe makes the system very reliable. More than 98% of PCR reaction can get successful amplification during the first try. It also works well on complicate templates.The Master Mix contains dye, and can directly run electrophoresis after PCR reaction.
Catalog No. NC3486594 Supplier 101 BIO Supplier No. W06901
This Master Mix contains Es Taq DNA Polymerase, PCR buffer, Mg2+, dNTP, PCR stabilizer and PCR enhancer. The concentration is 2x.Es Taq DNA Polymerase possesses 5´→3´ DNA polymerase and 5´→3´ exonuclease activity. The polymerase has the high amplification efficiency and low mismatching as Taq DNA polymerase and high fidelity of Pfu DNA polymerase. Es Taq Polymerase catalyzes the non-template directed addition of an adenine residue to the 3´-end of both strands of DNA molecules to make it suitable for T/A cloning. The amplification range of Es Taq is ~ 6 kb.This unique Master Mix recipe makes the system very reliable. More than 98% of PCR reaction can get successful amplification during the first try. It also works well on complicate templates.The Master Mix contains dye, and can directly run electrophoresis after PCR reaction.