Learn More
101 BIO 2x GoldStar Best PCR Master Mix (with dye), (1 mL, 100 rxn, 20 ul/rxn)
Supplier:  101 BIO W06551
Encompass
2x GoldStar Best PCR Master Mix (with dye) is a convenient premixed 2x concentrated solution for PCR which include GoldStar Best DNA Polymerase, PCR Buffer, Mg2+, dNTPs, PCR Stabilizer and PCR Intensifier.The polymerase has a 5´→3´ DNA polymerase and a 5´→3´ exonuclease activity. It also possesses 3´→5´ exonuclease (proofreading) activity. The polymerase has a higher amplification efficiency and lower mispairing rate than routine Taq DNA polymerase.The GoldStar Best is a high reliable hotstart polymerase, which activity is inhibited at ambient temperatures by the chemical modificaiton. This prevents the formation of misprimed products and primer dimers at ambient temperatures. GoldStar Best Polymerase is activated by a 10 minutes, 95°C incubation. Optimized buffer system promotes the amplification of target fragment with high fidelity, high specificity, high amplification efficiency and high sensitivity.
Catalog No. NC3486595