Learn More
Teknova 2X HBS (HEPES-Buffered Saline), pH 7.05. 250mL, Endotoxin Tested. Sterile. 4 pack.
SDP
Catalog No. 502745801
Encompass
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.
Catalog No. 50-274-5801 Supplier Teknova Supplier No. H17054PK
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.