Learn More
TRIBIOSCIENCE INC 2x HBS, pH 7.05 (Sterile, Tissue Culture Grade) 500 ML
SDP
Catalog No. NC3343207
Encompass
Tribioscience’s HBS (2x HEPES buffered saline) is ready to use for chemical transfection, including calcium phosphate transfection and purposes such as calcium transfection, calcium phosphate transfection, mammalian cell transfection, virus production, lentivirus and retrovirus transfection, and AAV transfection.
Catalog No. NC3343207 Supplier TRIBIOSCIENCE INC Supplier No. TBS5076
Tribioscience’s HBS (2x HEPES buffered saline) is ready to use for chemical transfection, including calcium phosphate transfection and purposes such as calcium transfection, calcium phosphate transfection, mammalian cell transfection, virus production, lentivirus and retrovirus transfection, and AAV transfection.