Learn More
Raybiotech Inc Lysis Buffer, 16 mL, 2X
Catalog No. NC1520075
Encompass_Preferred
16 mL of 2X Lysis Buffer. 2X Lysis Buffer should be diluted 2-fold with deionized or distilled water. Supplementation with protease or phosphatase inhibitors is recommended. You may use RayBiotech Protease Inhibitor Cocktail (AA-PI), RayBiotech Phosphatase Inhibitor Cocktail Set I (AA-PHI-I) or Phosphatase Inhibitor Cocktail Set II (AA-PHI-II), or the RayBiotech Phosphatase / Protease Inhibitor Set (AA-PIPHI). Alternatively, you can purchase inhibitor cocktails from another manufacturer and follow their specifications.
Catalog No. NC1520075 Supplier Raybiotech Inc Supplier No. AALYS16ML
16 mL of 2X Lysis Buffer. 2X Lysis Buffer should be diluted 2-fold with deionized or distilled water. Supplementation with protease or phosphatase inhibitors is recommended. You may use RayBiotech Protease Inhibitor Cocktail (AA-PI), RayBiotech Phosphatase Inhibitor Cocktail Set I (AA-PHI-I) or Phosphatase Inhibitor Cocktail Set II (AA-PHI-II), or the RayBiotech Phosphatase / Protease Inhibitor Set (AA-PIPHI). Alternatively, you can purchase inhibitor cocktails from another manufacturer and follow their specifications.