Learn More
Raybiotech Inc Lysis Buffer, 5 mL, 2X
Catalog No. NC0996662
Encompass_Preferred
5 mL of 2X Lysis Buffer. 2X Lysis Buffer should be diluted 2-fold with deionized or distilled water. Supplementation with protease or phosphatase inhibitors is recommended. You may use RayBiotech Protease Inhibitor Cocktail (AA-PI), RayBiotech Phosphatase Inhibitor Cocktail Set I (AA-PHI-I) or Phosphatase Inhibitor Cocktail Set II (AA-PHI-II), or the RayBiotech Phosphatase / Protease Inhibitor Set (AA-PIPHI). Alternatively, you can purchase inhibitor cocktails from another manufacturer and follow their specifications. For the first experiment, we recommend serial dilution testing, starting with a 5-fold dilution, to determine the optimal protein load for the assay. Optimal dilution depends on the abundance of target proteins and should be determined empirically.
Catalog No. NC0996662 Supplier Raybiotech Inc Supplier No. ELLYSIS
5 mL of 2X Lysis Buffer. 2X Lysis Buffer should be diluted 2-fold with deionized or distilled water. Supplementation with protease or phosphatase inhibitors is recommended. You may use RayBiotech Protease Inhibitor Cocktail (AA-PI), RayBiotech Phosphatase Inhibitor Cocktail Set I (AA-PHI-I) or Phosphatase Inhibitor Cocktail Set II (AA-PHI-II), or the RayBiotech Phosphatase / Protease Inhibitor Set (AA-PIPHI). Alternatively, you can purchase inhibitor cocktails from another manufacturer and follow their specifications. For the first experiment, we recommend serial dilution testing, starting with a 5-fold dilution, to determine the optimal protein load for the assay. Optimal dilution depends on the abundance of target proteins and should be determined empirically.