Learn More
Teknova 2X Mueller Hinton II Cation Adjusted Broth. 1000mL, Sterile. 2 pack.
SDP
Catalog No. 502387559
Encompass
Mueller-Hinton is a growth medium that is commonly used for antibiotic susceptibility testing, due to its moderate nutrient levels and uniformity. This makes it ideal for performing disk diffusion tests, which help determine effectiveness of antibiotics against bacteria.
Catalog No. 50-238-7559 Supplier Teknova Supplier No. M58662PK
Mueller-Hinton is a growth medium that is commonly used for antibiotic susceptibility testing, due to its moderate nutrient levels and uniformity. This makes it ideal for performing disk diffusion tests, which help determine effectiveness of antibiotics against bacteria.