Learn More
Norgen Biotek Corp 2X One-Step Rt-Pcr Mas 200Rxns
Supplier:  Norgen Biotek Corp 28114
Encompass_Preferred
Norgen’s 2X One-Step RT-PCR Master Mix is a ready-to-use solution that contains components required for RT-PCR amplification of RNA templates. The mix includes M-MuLV reverse transcriptase, Taq DNA polymerase, dNTPs, reaction buffer, MgCl2, KCl, and a PCR enhancer/stabilizer. The user needs only to add the template, the primer set and water to the Master Mix to set up the RT-PCR reaction. This convenient 2X One-Step RT-PCR Master Mix reduces the time required to set up PCR reactions and reduces the possibility of contamination, particularly when preparing large numbers of reactions. The optimized master mix allows for robust amplification of RNA templates with high yields of PCR products.Taq Source: An E. coli strain with a cloned Taq DNA Polymerase gene from Thermus aquaticus YT-1RT Source: An E. coli strain with a cloned reverse transcriptase gene from M-MuLV.
Catalog No. NC1810691