Learn More
Apexbio Technology LLC 2X hyperfusion plus master mix 10X1ml
SDP
Catalog No. 503107293
Encompass_Preferred
Our 2X Hyperfusion Plus Master Mix (With dye) product offers high fidelity strong amplification performance and faster extension speed simultaneously making it suitable for most PCR applications Hyperfusion Plus DNA Polymerase is created by fusing a Pyrococcus-like proofreading polymerase with a DNA binding domain providing increased fidelity and speed Hyperfusion Plus DNA Polymerase owns high fidelity 50 times lower error rate than Taq DNA Polymerase and 6 times lower error compared to Pyrococcus Furious DNA Polymerase Hyperfusion Plus DNA Polymerase exhibits both 5 to 3 polymerase activity and 3 to 5 exonuclease (proofreading) activity producing blunt-ended amplification products rather than A-tailed ends (commonly produced by Taq polymerase) Experimental results demonstrate that our Hyperfusion Plus polymerase can amplify genome fragments as long as 20 1 kb
Catalog No. 50-310-7293 Supplier Apexbio Technology LLC Supplier No. K111910
Our 2X Hyperfusion Plus Master Mix (With dye) product offers high fidelity strong amplification performance and faster extension speed simultaneously making it suitable for most PCR applications Hyperfusion Plus DNA Polymerase is created by fusing a Pyrococcus-like proofreading polymerase with a DNA binding domain providing increased fidelity and speed Hyperfusion Plus DNA Polymerase owns high fidelity 50 times lower error rate than Taq DNA Polymerase and 6 times lower error compared to Pyrococcus Furious DNA Polymerase Hyperfusion Plus DNA Polymerase exhibits both 5 to 3 polymerase activity and 3 to 5 exonuclease (proofreading) activity producing blunt-ended amplification products rather than A-tailed ends (commonly produced by Taq polymerase) Experimental results demonstrate that our Hyperfusion Plus polymerase can amplify genome fragments as long as 20 1 kb