Learn More
Apexbio Technology LLC 2X hyperfusion plus master mix (With dye) 10X1ml
SDP
Catalog No. 503107348
Encompass_Preferred
Our 2X Hyperfusion Plus Master Mix (With dye) product offers high fidelity strong amplification performance and faster extension speed simultaneously making it suitable for most PCR applications Hyperfusion Plus DNA Polymerase is created by fusing a Pyrococcus-like proofreading polymerase with a DNA binding domain providing increased fidelity and speed Hyperfusion Plus DNA Polymerase owns high fidelity 50 times lower error rate than Taq DNA Polymerase and 6 times lower error compared to Pyrococcus Furious DNA Polymerase Experimental results demonstrate that our Hyperfusion Plus polymerase can amplify genome fragments as long as 20 1 kb 2X Hyperfusion plus master mix (With dye) is a ready-to-use 2 premix solution containing polymerase dNTPs an already optimized buffer system and dye Our mix is ideal for any detection and the PCR product can be directly electrophoresed after the amplification without the need to add a loading buffer
Catalog No. 50-310-7348 Supplier Apexbio Technology LLC Supplier No. K111710
Our 2X Hyperfusion Plus Master Mix (With dye) product offers high fidelity strong amplification performance and faster extension speed simultaneously making it suitable for most PCR applications Hyperfusion Plus DNA Polymerase is created by fusing a Pyrococcus-like proofreading polymerase with a DNA binding domain providing increased fidelity and speed Hyperfusion Plus DNA Polymerase owns high fidelity 50 times lower error rate than Taq DNA Polymerase and 6 times lower error compared to Pyrococcus Furious DNA Polymerase Experimental results demonstrate that our Hyperfusion Plus polymerase can amplify genome fragments as long as 20 1 kb 2X Hyperfusion plus master mix (With dye) is a ready-to-use 2 premix solution containing polymerase dNTPs an already optimized buffer system and dye Our mix is ideal for any detection and the PCR product can be directly electrophoresed after the amplification without the need to add a loading buffer