Learn More
LAMDA BIOTECH INC 2X qPCR Green MasterMix
Catalog No. 502045774
Encompass_Preferred
2X qPCR Universal Green MasterMix is a qPCR master mix contains a EvaGreen fluorescent dye for SYBR Green Assays in quantitative, real-time PCR analysis. With the heat stable EvaGreen dye and new generation of DNA polymerase, this specially formulated qPCR master mix will significantly increase the processivity and sensitivity; reduce primer dimmers and non-specificity; and shorten the required total qPCR time. Without any modification, it can be directly used for different SYBR Green assays.
Catalog No. 50-204-5774 Supplier LAMDA BIOTECH INC Supplier No. QMXGREEN
2X qPCR Universal Green MasterMix is a qPCR master mix contains a EvaGreen fluorescent dye for SYBR Green Assays in quantitative, real-time PCR analysis. With the heat stable EvaGreen dye and new generation of DNA polymerase, this specially formulated qPCR master mix will significantly increase the processivity and sensitivity; reduce primer dimmers and non-specificity; and shorten the required total qPCR time. Without any modification, it can be directly used for different SYBR Green assays.