Learn More
LAMDA BIOTECH INC 2X qPCR TaqProbe MasterMix-25ml
Catalog No. 502045777
Encompass_Preferred
2X qPCR Universal TaqProbe MasterMix is a qPCR master mix designed for TaqMan Assays in quantitative, real-time PCR analysis. With the specially engineered DNA polymerase and its unique qPCR buffer system, this universal qPCR master mix offers superb performance in sensitivity and signal-to-noise ratio. Without any modifications, the 2X qPCR Universal TaqProbe MasterMix can be directly used for any TaqMan assays, such as SNP genotyping, gene expression analysis, microarray validation, and high throughput screening applications. The ROX reference dye is provided separately, making it more flexible and compatible with different qPCR machines.
Catalog No. 50-204-5777 Supplier LAMDA BIOTECH INC Supplier No. QMXTAQM25ML
2X qPCR Universal TaqProbe MasterMix is a qPCR master mix designed for TaqMan Assays in quantitative, real-time PCR analysis. With the specially engineered DNA polymerase and its unique qPCR buffer system, this universal qPCR master mix offers superb performance in sensitivity and signal-to-noise ratio. Without any modifications, the 2X qPCR Universal TaqProbe MasterMix can be directly used for any TaqMan assays, such as SNP genotyping, gene expression analysis, microarray validation, and high throughput screening applications. The ROX reference dye is provided separately, making it more flexible and compatible with different qPCR machines.