Learn More
Teknova 2X R2A Broth. 1000mL, Sterile. 4 pack.
SDP
Catalog No. 502387539
Encompass
0.05% Proteose Peptone #3,0.05% Tryptone,0.10% Casamino Acids,0.10% Yeast Extract,0.10% Cerelose Anhydrous Dextrose (Glucose),0.10% Sodium Pyruvate,0.06% Potassium Phosphate Dibasic,0.01% Magnesium Sulfate Heptahydrate
Catalog No. 50-238-7539 Supplier Teknova Supplier No. R10374PK
Add to Cart
Add to Cart
0.05% Proteose Peptone #3,0.05% Tryptone,0.10% Casamino Acids,0.10% Yeast Extract,0.10% Cerelose Anhydrous Dextrose (Glucose),0.10% Sodium Pyruvate,0.06% Potassium Phosphate Dibasic,0.01% Magnesium Sulfate Heptahydrate