Learn More
Laxco 2X Supplementary Objective for Z200PRO Series Stereo Microscope
GREENER_CHOICESDP
Catalog No. 12563724 Shop All Laxco Inc Products
Change view
Click to view available options
Type:
Auxiliary Lens
Catalog No. Type
12-563-724 Auxiliary Lens
1 options
Catalog No. 12-563-724 Supplier Laxco Supplier No. LMSPZ200PSL200
Add to Cart
Add to Cart
Enables users to expand their magnification range by two times on their existing LAXCO stereo microscopes. Now get a more magnified and clearer view of your samples with this field upgradable accesory.
  • Auxiliary Lens: Able to extend the working distance and field view of the microscope, this objective lens will simplify your working.
  • Clear Viewing: 2X magnification enables a clearer viewing on the object, which will also bring you more precision in the observation.
  • Perfect Match: Suits perfectly for LAXCO Z200PRO series stereo microscope
  • Good Material: Adopting premium alloy material, not likely to break, and also has excellent durability for long term using.
  • Easy to Install: Compact size and a light weight, the convenient installation can be quickly completed within minutes.
Specifications
Type Auxiliary Lens
Autoclavable Not Autoclavable
Sterility Sterile