Learn More
TRIBIOSCIENCE INC 2x Sybr qPCR Super Mix, ROX, 2 x 1.25 ML
SDP
Catalog No. NC3834041
Encompass
Tribioscience 2x Fast Sybr Green qPCR Master Mix, ROX, 2 x 1.25 ML provides a convenient choice suitable for different qPCR instruments. It is a 2x concentrated premix containing Taq DNA polymerase, dNTPs, Mg2+, SYBR Green I dye (detection), ROX reference dye and PCR buffer components.
Catalog No. NC3834041 Supplier TRIBIOSCIENCE INC Supplier No. TBS4001R2.5
Tribioscience 2x Fast Sybr Green qPCR Master Mix, ROX, 2 x 1.25 ML provides a convenient choice suitable for different qPCR instruments. It is a 2x concentrated premix containing Taq DNA polymerase, dNTPs, Mg2+, SYBR Green I dye (detection), ROX reference dye and PCR buffer components.