Learn More
LAMDA BIOTECH INC 2X YT
Catalog No. 502045828
Encompass_Preferred
Premixed 2xYT Broth powder: weigh it out once, put it in the flask or bottle, add the appropriate volume of water and autoclave as usual.
Catalog No. 50-204-5828 Supplier LAMDA BIOTECH INC Supplier No. C132
Premixed 2xYT Broth powder: weigh it out once, put it in the flask or bottle, add the appropriate volume of water and autoclave as usual.