Learn More
Sigma Aldrich Fine Chemicals Biosciences 2x YT medium Powder microb250G
Catalog No. 502900433
Encompass
2xYT is a standard microbial growth medium used for the cultivation of E. coli and M13 bacteriophage. This nutrient-rich microbial broth contains peptides amino acids and water-soluble vitamins in a low-salt formulation.
Catalog No. 50-290-0433 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Y2377250G
2xYT is a standard microbial growth medium used for the cultivation of E. coli and M13 bacteriophage. This nutrient-rich microbial broth contains peptides amino acids and water-soluble vitamins in a low-salt formulation.