Learn More
Roche Diagnostics KAPA Express Extract Kit | Standard Kit, 200u
Supplier:  Roche Diagnostics 07961600001
Encompass
KAPA Express Extract Kit is a novel thermostable protease and buffer system that allows for the extraction of PCR-ready DNA in as little as 15 minutes. DNA extractions are conveniently performed in a single-tube thereby greatly reducing the risk of sample loss and contamination. *This item is non returnable.
Catalog No. 50-196-5275