Learn More
MSC Lift-All Endless Web Sling, Type 5, 12 ft, 2 in, Nylon
Catalog No. 5000534367
Encompass
This Lift-All endless web sling is a Type 5 design, crafted from durable nylon material. It measures 12 feet in length and 2 inches in width, featuring a 2-ply construction for enhanced strength. With a vertical capacity of 12400 pounds, a choker capacity of 9900 pounds, and a basket capacity of 24800 pounds, this abrasion-resistant sling is designed for robust lifting applications. It meets WSTDA, ASME B30.9, and OSHA 1910.184 standards, making it suitable for various industrial environments.
  • Abrasion-resistant construction for extended lifespan.
  • Conforms to WSTDA standards for quality assurance.
  • Meets ASME B30.9 safety guidelines.
  • Complies with OSHA 1910.184 regulations for workplace safety.
  • Yellow color for high visibility.
  • Suitable for operations up to 200°F.
Catalog No. 50-005-34367 Supplier MSC Supplier No. 87843983
This Lift-All endless web sling is a Type 5 design, crafted from durable nylon material. It measures 12 feet in length and 2 inches in width, featuring a 2-ply construction for enhanced strength. With a vertical capacity of 12400 pounds, a choker capacity of 9900 pounds, and a basket capacity of 24800 pounds, this abrasion-resistant sling is designed for robust lifting applications. It meets WSTDA, ASME B30.9, and OSHA 1910.184 standards, making it suitable for various industrial environments.
  • Abrasion-resistant construction for extended lifespan.
  • Conforms to WSTDA standards for quality assurance.
  • Meets ASME B30.9 safety guidelines.
  • Complies with OSHA 1910.184 regulations for workplace safety.
  • Yellow color for high visibility.
  • Suitable for operations up to 200°F.