Learn More
Biosurface Technologies Corp Borosilicate Glass Square Coupon, 2 inch x 2 inch
Catalog No. NC0651024
Encompass
This 2 inch by 2 inch square coupon is manufactured from borosilicate glass, and is designed for biofilm or biofilm-related studies in a laboratory environment. This item has a minimum quantity of 5 per supplier requirements.
Catalog No. NC0651024 Supplier Biosurface Technologies Corp Supplier No. BST 5050GL
This 2 inch by 2 inch square coupon is manufactured from borosilicate glass, and is designed for biofilm or biofilm-related studies in a laboratory environment. This item has a minimum quantity of 5 per supplier requirements.