Learn More
Biosurface Technologies Corp Polypropylene Square Coupon, 2 inch x 2 inch
Catalog No. NC1721124
Encompass
This 2 inch by 2 inch square coupon is manufactured from polypropylene, and is designed for biofilm or biofilm-related studies in a laboratory environment. This item has a minimum quantity of 6 per supplier requirements.
Catalog No. NC1721124 Supplier Biosurface Technologies Corp Supplier No. BST 5050PP
This 2 inch by 2 inch square coupon is manufactured from polypropylene, and is designed for biofilm or biofilm-related studies in a laboratory environment. This item has a minimum quantity of 6 per supplier requirements.