Learn More
Uline Anti-Static Mat with Cord - 2 x 3,1 EA
Catalog No. NC0453368
Encompass
Eliminate electrical shock damage. Foam vinyl mat reduces muscle strain. 10 grounding cord included. Meets MIL-HDBK MIL-STD-1686 RTG109-10 Ohms EOSESD 71-1991. Grounding Cord: Ships unattached, Snaps on and off, Must be fastened to a grounded wall outlet. Mat can be cut without affecting anti-static properties. Not for use under casters or chair wheels.
Catalog No. NC0453368 Supplier Uline Supplier No. H894
Eliminate electrical shock damage. Foam vinyl mat reduces muscle strain. 10 grounding cord included. Meets MIL-HDBK MIL-STD-1686 RTG109-10 Ohms EOSESD 71-1991. Grounding Cord: Ships unattached, Snaps on and off, Must be fastened to a grounded wall outlet. Mat can be cut without affecting anti-static properties. Not for use under casters or chair wheels.