Learn More
MSC Lift-All Web Sling, 2-Ply Flat Eye & Eye, Polyester, 2 in. x 4 ft.
Catalog No. 5000534146
Encompass
This 2-ply, flat eye & eye polyester web sling is designed as an economical option for light to moderate duty applications and less frequent use. It incorporates essential features for safety and durability, promoting reliable load handling and extended service life.
  • Red core yarn warning system aids in the inspection process by visually indicating damage.
  • Tuff-Tag™ provides serial-numbered identification for traceability and required OSHA information for the life of the sling.
  • Economical option for light to moderate duty applications and less frequent use.
  • Yellow treatment provides improved abrasion resistance and extends sling life.
  • Polyester material stretches less, offering better load control and reduced abrasion.
Catalog No. 50-005-34146 Supplier MSC Supplier No. 01136803
This 2-ply, flat eye & eye polyester web sling is designed as an economical option for light to moderate duty applications and less frequent use. It incorporates essential features for safety and durability, promoting reliable load handling and extended service life.
  • Red core yarn warning system aids in the inspection process by visually indicating damage.
  • Tuff-Tag™ provides serial-numbered identification for traceability and required OSHA information for the life of the sling.
  • Economical option for light to moderate duty applications and less frequent use.
  • Yellow treatment provides improved abrasion resistance and extends sling life.
  • Polyester material stretches less, offering better load control and reduced abrasion.