Learn More
Apexbio Technology LLC 2SYBR Green qPCR Master Mix, 500rxn (20 ul reaction)
SDP
Catalog No. 501998223
Encompass_Preferred
2SYBR Green qPCR Master Mix is a ready-to-use 2X master mix optimized for highly specific and sensitive real-time quantitative PCR using SYBR Green I and Hot Start Taq DNA Polymerase. SYBR Green I is a fluorescent double-stranded DNA binding dye that tracks double-stranded DNA amplified in PCR. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8223 Supplier Apexbio Technology LLC Supplier No. K10705
2SYBR Green qPCR Master Mix is a ready-to-use 2X master mix optimized for highly specific and sensitive real-time quantitative PCR using SYBR Green I and Hot Start Taq DNA Polymerase. SYBR Green I is a fluorescent double-stranded DNA binding dye that tracks double-stranded DNA amplified in PCR. Other sizes are also available. Please inqury us for quote.