Learn More
Bioland Scientific LLC 2x Taq PCR Premix (5x1 ml, 400 RXN, Green)
SDP
Catalog No. NC1837689
Encompass_Preferred
An optimized ready-to-use solution containing Taq DNA polymerase, standard Taq reaction buffer, dNTPs, tracking dyes and stabilizers. It is ideally suited to routine PCR applications such as sub-cloning, colony screening and genotyping. It can amplify up to 4 kb from complex genomic DNA or up to 5 kb from lambda DNA.
Catalog No. NC1837689 Supplier Bioland Scientific LLC Supplier No. TP0101
An optimized ready-to-use solution containing Taq DNA polymerase, standard Taq reaction buffer, dNTPs, tracking dyes and stabilizers. It is ideally suited to routine PCR applications such as sub-cloning, colony screening and genotyping. It can amplify up to 4 kb from complex genomic DNA or up to 5 kb from lambda DNA.