Learn More
MSC MSC Double-Faced Corrugated Sheet, 36 x 36 in, Kraft, 5/Box
Catalog No. 501919669
Encompass
This corrugated sheet is designed for use as a protective layer on pallets. It features a double-faced construction and comes in a kraft color, providing a reliable solution for various packaging and storage needs.
  • Suitable for use as layering pads to distribute weight
  • Helps keep products dust-free during storage
  • Prevents damage from strapping and forklifts
  • Sold in bundle quantities
Catalog No. 50-191-9669 Supplier MSC Supplier No. 39592852
This corrugated sheet is designed for use as a protective layer on pallets. It features a double-faced construction and comes in a kraft color, providing a reliable solution for various packaging and storage needs.
  • Suitable for use as layering pads to distribute weight
  • Helps keep products dust-free during storage
  • Prevents damage from strapping and forklifts
  • Sold in bundle quantities