Learn More
Teknova 2XYT Agar. 1Liter Foil Pouch. 10 pack
SDP
Catalog No. 502388132
Encompass
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.8% Agar
Catalog No. 50-238-8132 Supplier Teknova Supplier No. Y0230
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.8% Agar