Learn More
Teknova 2XYT Agar. 24.5g in Foil Pouches. 10 Pouches per Pack. Reconstitute in 500mL.
SDP
Catalog No. 502388133
Encompass
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.8% Agar
Catalog No. 50-238-8133 Supplier Teknova Supplier No. Y0235
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.8% Agar