Learn More
Teknova 2XYT Agar Pates with 2% Glucose, Carbenicillin-100. 86x128mm, 20 plates/sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387458
Encompass
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar, 2.0% Cerelose Anhydrous Dextrose (Glucose), 100g/mL Carbenicillin
Catalog No. 50-238-7458 Supplier Teknova Supplier No. Y23908PK
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar, 2.0% Cerelose Anhydrous Dextrose (Glucose), 100g/mL Carbenicillin