Learn More
Teknova 2XYT Agar Plates. 100mm, 20 plates/sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387519
Encompass
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar
Catalog No. 50-238-7519 Supplier Teknova Supplier No. Y42104PK
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar