Learn More
Teknova 2XYT Agar Plates with 2% Glucose and Ampicillin-100. 245mm, 4 Plates per Sleeve, Sterile. 8 Pack.
SDP
Catalog No. 502387889
Encompass
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 2.0% Agar,2.0% Cerelose Anhydrous Dextrose (Glucose), 100g/mL Ampicillin Sodium Salt
Catalog No. 50-238-7889 Supplier Teknova Supplier No. Y42928PK
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 2.0% Agar,2.0% Cerelose Anhydrous Dextrose (Glucose), 100g/mL Ampicillin Sodium Salt