Learn More
Teknova 2XYT Agar Plates with Carbenicillin-50. 100mm, 20 Plates per Sleeves, Sterile. 8 Pack.
SDP
Catalog No. 502387792
Encompass
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar,50g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-7792 Supplier Teknova Supplier No. Y42308PK
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar,50g/mL Carbenicillin Disodium Salt