Learn More
Teknova 2XYT Agar Plates with 2% Glucose and Carbenicillin-100. 245mm Plate, 4 Plates per Sleeve, Sterile. 4 Pack.
SDP
Supplier:  Teknova Y62604PK
Encompass
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 2.0% Glucose, 1.5% Agar, 100g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-7620