Learn More
Teknova 2XYT Agar Plates with 2% Glucose and Carbenicillin-100. 245mm Nunc Low Profile Plates, 300mL, Sterile. 4 Pack.
SDP
Catalog No. 502387506
Encompass
1.6% Tryptone, 1.0% Yeast Extract,0.5% Sodium Chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt, 2.0% Glucose
Catalog No. 50-238-7506 Supplier Teknova Supplier No. Y64924PK
1.6% Tryptone, 1.0% Yeast Extract,0.5% Sodium Chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt, 2.0% Glucose