Learn More
Teknova 2XYT Agar Plates with 2% Glucose, Carbenicillin-100. 100mm Plates, 20 Plates per Sleeve, Sterile. 4 Pack.
SDP
Catalog No. 502387631
Encompass
1.6% Tryptone, 1.0% Yeast Extract,0.5% Sodium Chloride, 1.5% Agar,2.0% Glucose, 100g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-7631 Supplier Teknova Supplier No. Y42074PK
1.6% Tryptone, 1.0% Yeast Extract,0.5% Sodium Chloride, 1.5% Agar,2.0% Glucose, 100g/mL Carbenicillin Disodium Salt