Learn More
Teknova 2XYT Agar Plates with 2% Glucose, Amp-100, Tet-20. 100mm, 20 Plates/Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387580
Encompass
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar, 2.0% Cerelose Anhydrous Dextrose (Glucose), 100g/mL Ampicillin Sodium Salt,20g/mL Tetracycline Hydrochloride
Catalog No. 50-238-7580 Supplier Teknova Supplier No. Y42278PK
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar, 2.0% Cerelose Anhydrous Dextrose (Glucose), 100g/mL Ampicillin Sodium Salt,20g/mL Tetracycline Hydrochloride