Learn More
Teknova 2XYT Agar Plates with 2% Glucose and Kanamycin-50. 100mm plates, 20 Plates per Sleeve, Sterile. 4 Pack.
SDP
Catalog No. 502387799
Encompass
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar, 2.0% Cerelose Anhydrous Dextrose (Glucose), 50g/mL Kanamycin
Catalog No. 50-238-7799 Supplier Teknova Supplier No. Y13204PK
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar, 2.0% Cerelose Anhydrous Dextrose (Glucose), 50g/mL Kanamycin