Learn More
Teknova 2XYT Agar Plates with 2% Glucose, Carb-100, and Tet-20. 150mm Plates, Sterile. 4 pack.
SDP
Catalog No. 502387346
Encompass
1.6% Tryptone, 1.0% Yeast extract, 0.5% Sodium chloride, 1.5% Agar, 2.0% Cerelose Anhydrous Dextrose (Glucose), 100g/mL Carbenicillin, 20g/mL Tetracycline Hydrochloride
Catalog No. 50-238-7346 Supplier Teknova Supplier No. Y24054PK
1.6% Tryptone, 1.0% Yeast extract, 0.5% Sodium chloride, 1.5% Agar, 2.0% Cerelose Anhydrous Dextrose (Glucose), 100g/mL Carbenicillin, 20g/mL Tetracycline Hydrochloride