Learn More
Teknova 2XYT Agar Plates with 2% Glucose and Carbenicillin-100.150mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502388098
Encompass
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 2.0% Glucose, 1.5% Agar, 100g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-8098 Supplier Teknova Supplier No. Y43914PK
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 2.0% Glucose, 1.5% Agar, 100g/mL Carbenicillin Disodium Salt