Learn More
Teknova 2XYT Agar Plates with Carbenicillin-100 ,100mm Plates, 20 Plates/Sleeve. Sterile. 4 pack.
SDP
Catalog No. 502387339
Encompass
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-7339 Supplier Teknova Supplier No. Y43934PK
1.6% Tryptone, 1.0% Yeast Extract, 0.5% Sodium Chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt